Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMP-9 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3879SP
This item is not returnable.
View return policy
Description
BMP-9 Polyclonal specifically detects BMP-9 in Human samples. It is validated for Western Blot.Specifications
BMP-9 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP9, BMP9BMP-9Bone morphogenetic protein 9, GDF2, GDF-2, growth differentiation factor 2, growth/differentiation factor 2 | |
Goat | |
Affinity Purified | |
RUO | |
2658 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q9UK05 | |
GDF2 | |
Chinese hamster ovary cell line CHO-derived recombinant human BMP-9 Propeptide Lys23-Arg319 Accession # Q9UK05 | |
25 μg | |
Primary | |
Detects human BMP-9 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with mature recombinant human BMP-9 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction