Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMPR-IB/ALK-6 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5051U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5051U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5051U100 Supplier R&D Systems Supplier No. FAB5051U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BMPR-IB/ALK-6 Monoclonal specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BMPR-IB/ALK-6
Applications Flow Cytometry
Classification Monoclonal
Clone 477914
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O00238
Gene Alias activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR1B, BMPR-1B, BMPRIB, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase
Gene Symbols BMPR1B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BMPR-IB/ALK-6 Lys14-Arg126 Accession # O00238
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 658
Test Specificity Detects human BMPR-IB/ALK-6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse BMPR-IB is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.