Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BMPR-IB/ALK-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5051
This item is not returnable.
View return policy
Description
BMPR-IB/ALK-6 Monoclonal specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
BMPR-IB/ALK-6 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
O00238 | |
BMPR1B | |
Mouse myeloma cell line NS0-derived recombinant human BMPR-IB/ALK-6 Lys14-Arg126 Accession # O00238 | |
100 μg | |
Primary | |
Detects human BMPR-IB/ALK-6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse BMPR-IB is observed. | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
477914 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR1B, BMPR-1B, BMPRIB, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
658 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction