Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BOC Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2036
This item is not returnable.
View return policy
Description
BOC Polyclonal specifically detects BOC in Human samples. It is validated for Western Blot.Specifications
BOC | |
Polyclonal | |
Unconjugated | |
Q9BWV1 | |
BOC | |
Mouse myeloma cell line NS0-derived recombinant human BOC Asp31-Asp852 Accession # Q9BWV1 | |
100 μg | |
Primary | |
Detects human BOC in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse BOC is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Boc homolog (mouse), brother of CDO, Protein BOC | |
Goat | |
Affinity Purified | |
RUO | |
91653 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction