Learn More
Specifications
Specifications
| Assay Range | 0.094 - 6 ng/mL (example only; lot dependent) |
| Assay Sensitivity | 64 pg/mL (example only; lot dependent) |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Culture, Plasma, Serum |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3035 |
| Quantity | 1 Kit |
| Storage Requirements | Storage is content dependent. |
| Synonym | cytoplasmic, EC 6.1.1.21, FLJ20491, histidine-tRNA ligase, histidyl-tRNA synthetase, histidyl-tRNA synthetase, cytoplasmic |
| Target | Human, Bovine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
