Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BPI Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC74681V100UG
This item is not returnable.
View return policy
Description
BPI Monoclonal specifically detects BPI in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
BPI | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, BPIFD1, CAP 57, rBPI | |
Human embryonic kidney cell line HEK-293-derived transfected with human BPI, Val32-Lys487, Accession # P17213 | |
100 μg | |
671 | |
Human | |
Supernatant |
Flow Cytometry | |
971526 | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
P17213 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human BPI in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction