Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Bradykinin RB2/BDKRB2 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9434V100UG
This item is not returnable.
View return policy
Description
Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry.Specifications
Bradykinin RB2/BDKRB2 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088 | |
NS0 mouse myeloma cell line transfected with human RB2/BDKRB2, Met1-Gln391, Accession No. P30411 | |
Primary | |
Detects human RB2/BDKRB2 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Flow Cytometry | |
471902 | |
Flow Cytometry 0.25-1 μg/106 cells | |
P30411 | |
Mouse | |
100 μg | |
624 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction