Learn More
Human Bradykinin RB2/BDKRB2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Bradykinin RB2/BDKRB2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 471902 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide |
| Gene Accession No. | P30411 |
| Gene Alias | B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.