Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Bradykinin RB2/BDKRB2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB47251100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB47251100 100 μg
MAB47251SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB47251100 Supplier R&D Systems Supplier No. MAB47251100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bradykinin RB2/BDKRB2 Monoclonal specifically detects Bradykinin RB2/BDKRB2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen Bradykinin RB2/BDKRB2
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 471916
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P30411
Gene Alias B2 bradykinin receptor, B2R, BDKRB2, BK2, BK-2, BK-2 receptor, BKR2, bradykinin receptor B2, BradykininRB2, BRB2, DKFZp686O088
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2, Met1-Gln391, Accession No. P30411
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 624.0
Test Specificity Detects human Bradykinin RB2/BDKRB2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.