Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Brain Membrane Tissue Lysate (Adult Membrane Alzheimer's)
SDP

Catalog No. NBL126447 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBL126447 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBL126447 Supplier R&D Systems™ Supplier No. NBL126447
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

The Human Brain Membrane Tissue Lysate (Adult Membrane Alzheimer's) is derived from human.

Specifications

Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Membrane Protein - Alzheimers Disease: Brain: Temporal Lobe
Cellular Fraction Membrane
Tissue Condition Alzheimer's
Cross Reactivity Human
Host Species Human
Product Type Human Brain Membrane Tissue Lysate (Adult Alzheimer's)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Product Line Novus
Protein State Native
Tissue Brain
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.