Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Brevican Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB40091SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB40091SP 25 μg
MAB40091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB40091SP Supplier R&D Systems Supplier No. MAB40091SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Brevican Monoclonal specifically detects Brevican in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Brevican
Applications Immunocytochemistry
Classification Monoclonal
Clone 450616
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias ALPBRE, BCAN, BEHAB, BEHABbrevican core protein, Brain-enriched hyaluronan-binding protein, brevican, Chondroitin sulfate proteoglycan 7, chondroitin sulfate proteoglycan BEHAB, chondroitin sulfate proteoglycan BEHAB/brevican, CSPG7, CSPG7brevican proteoglycan, MGC13038
Gene Symbols BCAN
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Brevican isoform 1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63827
Test Specificity Detects human Brevican in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.