Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BRIP1/FANCJ Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6496SP
This item is not returnable.
View return policy
Description
BRIP1/FANCJ Monoclonal specifically detects BRIP1/FANCJ in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BRIP1/FANCJ | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9BX63 | |
BRIP1 | |
E. coli-derived recombinant human BRIP1/FANCJ Ser2-Arg160 Accession # Q9BX63 | |
25 μg | |
Primary | |
Detects human BRIP1/FANCJ in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) FANCA, D2, E, F, G, I, L, M, or recombinant mouse BRIP1/FANCJ is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
652747 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ATP-dependent RNA helicase BRIP1, BACH1, BACH1BRCA1/BRCA2-associated helicase 1, BRCA1 interacting protein C-terminal helicase 1, BRCA1-associated C-terminal helicase 1, BRCA1-binding helicase-like protein BACH1, BRCA1-interacting protein 1, BRCA1-interacting protein C-terminal helicase 1, EC 3.6.1, EC 3.6.4.13, FACJ, FANCJ, FANCJFanconi anemia group J protein, FLJ90232, MGC126521, MGC126523, OF, Protein FACJ | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
83990 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction