Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BRIP1/FANCJ Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6496
This item is not returnable.
View return policy
Description
BRIP1/FANCJ Monoclonal specifically detects BRIP1/FANCJ in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BRIP1/FANCJ | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9BX63 | |
BRIP1 | |
E. coli-derived recombinant human BRIP1/FANCJ Ser2-Arg160 Accession # Q9BX63 | |
100 μg | |
Primary | |
Detects human BRIP1/FANCJ in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) FANCA, D2, E, F, G, I, L, M, or recombinant mouse BRIP1/FANCJ is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
652747 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ATP-dependent RNA helicase BRIP1, BACH1, BACH1BRCA1/BRCA2-associated helicase 1, BRCA1 interacting protein C-terminal helicase 1, BRCA1-associated C-terminal helicase 1, BRCA1-binding helicase-like protein BACH1, BRCA1-interacting protein 1, BRCA1-interacting protein C-terminal helicase 1, EC 3.6.1, EC 3.6.4.13, FACJ, FANCJ, FANCJFanconi anemia group J protein, FLJ90232, MGC126521, MGC126523, OF, Protein FACJ | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
83990 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction