Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BTLA Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB33542U100UG
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry.Specifications
BTLA/CD272 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2161A | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAP44003 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
151888 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction