Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BTLA Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

Supplier:  R&D Systems FAB33542T100UG

Catalog No. FAB33542T10


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

BTLA/CD272
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003
100 μg
Primary
Human
Purified
Flow Cytometry
2161A
Flow Cytometry 0.25-1 ug/10^6 cells
AAP44003
Rabbit
Protein A or G purified from cell culture supernatant
RUO
151888
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.