Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BTLA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB33542SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB33542SP 25 μg
MAB33542100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB33542SP Supplier R&D Systems Supplier No. MAB33542SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen BTLA/CD272
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2161A
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. AAP44003
Gene Alias B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151888
Test Specificity Detects human BTLA in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.