Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BTN1A1/Butyrophilin Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB84671G100UG
This item is not returnable.
View return policy
Description
BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry.Specifications
BTN1A1/Butyrophilin | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), BTN, BTN1A1, butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin Ala27-Arg242 Accession # Q13410 | |
100 μg | |
696 | |
Human | |
Supernatant |
Flow Cytometry | |
2151B | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q13410 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human BTN1A1/Butyrophilin in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction