Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BTN1A1/Butyrophilin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB84671SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB84671SP 25 μg
MAB84671100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB84671SP Supplier R&D Systems Supplier No. MAB84671SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen BTN1A1/Butyrophilin
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 2151B
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q13410
Gene Alias BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), BTN, BTN1A1, butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human BTN1A1/Butyrophilin Ala27-Arg242 Accession # Q13410
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 696
Test Specificity Detects human BTN1A1/Butyrophilin in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.