Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BTN2A2/Butyrophilin 2A2 Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF8645SP
This item is not returnable.
View return policy
Description
BTN2A2/Butyrophilin 2 Polyclonal specifically detects BTN2A2/Butyrophilin 2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
BTN2A2/Butyrophilin 2 | |
Polyclonal | |
Western Blot 1:1000 dilution, Immunohistochemistry 1:50 dilution | |
Q8WVV5 | |
BTN2A2 | |
E. coli-derived recombinant human BTN2A2/Butyrophilin 2A2 Phe34-Val237 Accession # Q8WVV5 | |
25 μL | |
Primary | |
Detects human BTN2A2/Butyrophilin 2A2 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse BTN2A2 is observed, less than 5% cross-reactivity with recombinant human (rh) BTN2A1 and rhBTN3A2 is observed, and less than 1% cross-reactivity with rhBTN1A1 is observed. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
BTF2, BTF2BT2.2butyrophilin 2, BTN2, Butyrophilin 2, butyrophilin subfamily 2 member A2, butyrophilin, subfamily 2, member A2, FLJ41908 | |
Rabbit | |
Affinity Purified | |
RUO | |
10385 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction