Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BTN2A2/Butyrophilin 2A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8645 Supplier R&D Systems Supplier No. AF8645
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BTN2A2/Butyrophilin 2 Polyclonal specifically detects BTN2A2/Butyrophilin 2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen BTN2A2/Butyrophilin 2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:50 dilution
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. Q8WVV5
Gene Alias BTF2, BTF2BT2.2butyrophilin 2, BTN2, Butyrophilin 2, butyrophilin subfamily 2 member A2, butyrophilin, subfamily 2, member A2, FLJ41908
Gene Symbols BTN2A2
Host Species Rabbit
Immunogen E. coli-derived recombinant human BTN2A2/Butyrophilin 2A2 Phe34-Val237 Accession # Q8WVV5
Purification Method Antigen Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10385
Test Specificity Detects human BTN2A2/Butyrophilin 2A2 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse BTN2A2 is observed, less than 5% cross-reactivity with recombinant human (rh) BTN2A1 and rhBTN3A2 is observed, and less than 1% cross-reactivity with rhBTN1A1 is observed.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.