Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BTN3A1/2/3 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7136P025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7136P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7136P025 Supplier R&D Systems Supplier No. FAB7136P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BTN3A1/2/3 Monoclonal specifically detects BTN3A1/2/3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BTN3A1/2/3
Applications Flow Cytometry
Classification Monoclonal
Clone 849203
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. O00481
Gene Alias BT3.1, BTF5, BTN3A1, butyrophilin, subfamily 3, member A1, CD277, dJ45P21.3 (butyrophilin, subfamily 3, member A1)
Gene Symbols BTN3A1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD277/BTN3A1, Met1-Gly254, Accession # O00481
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11119
Test Specificity Detects human BTN3A1, BTN3A2 and BTN3A3 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.