Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BTNL8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9359SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9359SP 25 μg
MAB9359100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9359SP Supplier R&D Systems Supplier No. MAB9359SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BTNL8 Monoclonal specifically detects BTNL8 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry.

Specifications

Antigen BTNL8
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 2187B
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Simple Western 50 μg/mL, Flow Cytometry 0.25 μg/5 x 10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q6UX41-1
Gene Alias BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK-293-derived Human BTNL8 Gln18-Lys238 Accession # Q6UX41-1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 79908
Test Specificity Detects human BTNL8 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.