Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human c-Maf Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8227SP
This item is not returnable.
View return policy
Description
c-Maf Monoclonal specifically detects c-Maf in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
c-Maf | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
O75444 | |
MAF | |
E. coli-derived recombinant human c-Maf Met1-Phe372 Accession # O75444 | |
25 μg | |
Primary | |
Detects human c-Maf in direct ELISA and Western Blot. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
883926 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, CCA4, cMaf, c-MAF, c-maf proto-oncogene, MAF2, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4094 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction