Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human c-Myc Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB36961100
This item is not returnable.
View return policy
Description
c-Myc Monoclonal specifically detects c-Myc in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated.Specifications
| c-Myc | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, cMyc, c-Myc, MYC, myc proto-oncogene protein, Myc2, MYCC, myc-related translation/localization regulatory factor, Niard, Nird, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| E.coli-derived recombinant human c-Myc, Arg66-Asp201, Accession No. P01106 | |
| 100 μg | |
| 4609.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, Immunoassay | |
| 2270A | |
| Western Blot 1 μg;/mL, Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 3-25 μg;/mL, CyTOF-ready, Knockout Validated | |
| P01106 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human c-Myc in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction