Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human c-Myc PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC3696C
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3696C 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3696C Supplier R&D Systems Supplier No. IC3696C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

c-Myc Monoclonal specifically detects c-Myc in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen c-Myc
Applications Flow Cytometry
Classification Monoclonal
Clone 90000000000
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P01106
Gene Alias avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, cMyc, c-Myc, MYC, myc proto-oncogene protein, Myc2, MYCC, myc-related translation/localization regulatory factor, Niard, Nird, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Gene Symbols MYC
Host Species Mouse
Immunogen C-terminal region peptide of human c-Myc Ala408-Ala439 Accession # P01106
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4609
Test Specificity Detects endogenous human c-Myc and c-Myc tagged proteins in Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.