Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human C-Reactive Protein/CRP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$239.50 - $2140.00
Specifications
Antigen | C-Reactive Protein/CRP |
---|---|
Clone | 232007 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17071SP
![]() |
R&D Systems
MAB17071SP |
25 μg |
Each for $239.50
|
|
|||||
MAB17071100
![]() |
R&D Systems
MAB17071100 |
100 μg |
Each for $602.50
|
|
|||||
MAB17071500
![]() |
R&D Systems
MAB17071500 |
500 μg |
Each for $2,140.00
|
|
|||||
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
C-Reactive Protein/CRP | |
LYOPH | |
ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
CRP | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
232007 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P02741 | |
1401 | |
Human plasma-derived CRP Accession # P02741 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CRP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title