Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human C-Reactive Protein/CRP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB17071RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB17071RSP 25 μg
MAB1771R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB17071RSP Supplier R&D Systems Supplier No. MAB17071RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen C-Reactive Protein/CRP
Applications ELISA
Classification Monoclonal
Clone 232007R
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P02741
Gene Alias C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Host Species Mouse
Immunogen Human plasma-derived CRP Accession # P02741
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1401
Test Specificity Detects human CRP in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.