Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human C-Reactive Protein/CRP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | C-ReactiveProtein/CRP |
---|---|
Clone | 987312 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB17072SP
![]() |
R&D Systems
MAB17072SP |
25 μg |
Each for $200.50
|
|
|||||
MAB17072100
![]() |
R&D Systems
MAB17072100 |
100 μg |
Each for $514.50
|
|
|||||
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA.Specifications
C-ReactiveProtein/CRP | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P02741 | |
1401 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
987312 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
Mouse myeloma cell line NS0-derived recombinant human C-Reactive Protein/CRP Phe17-Pro224 Accession # P02741 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human C-Reactive Protein/CRP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title