Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human C-Reactive Protein/CRP Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems BAM17072
This item is not returnable.
View return policy
Description
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).Specifications
C-Reactive Protein/CRP | |
Monoclonal | |
LYOPH | |
ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1401 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
ELISA | |
232024 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
CRP | |
Human plasma-derived CRP | |
50 μg | |
Primary | |
Detects rhCRP in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction