Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human C-Reactive Protein/CRP Quantikine ELISA Kit

Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
Specifications
Specifications
Specificity | Natural and recombinant human CRP. |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 1401 |
Quantity | 1 Kit |
Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Synonym | C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 |
Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction