Learn More
Description
Specifications
Specifications
| Antigen | GPR15L |
| Applications | CyTOF, ELISA, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 1020904 |
| Conjugate | Unconjugated |
| Dilution | ELISA, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. |
| Gene Alias | AP57, AP-57, C10orf99 chromosome 10 open reading frame 99, GPR15L, UNQ1833 |
| Host Species | Mouse |
| Immunogen | E. coli-derived human C10orf99 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

