Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human C10orf99 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | GPR15L |
---|---|
Clone | 1020903 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10416SP
![]() |
R&D Systems
MAB10416SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10416100
![]() |
R&D Systems
MAB10416100 |
100 μg |
Each for $432.50
|
|
|||||
Description
GPR15L Monoclonal specifically detects GPR15L in Human samples. It is validated for ELISA.Specifications
GPR15L | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
387695 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1020903 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
AP57, AP-57, C10orf99 chromosome 10 open reading frame 99, GPR15L, UNQ1833 | |
E. coli-derived human C10orf99 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title