Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C14orf21 Full-length ORF (AAH25332.1, 1 a.a. - 536 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89937186
Click to view available options
:
2μg

Used for AP, Array, ELISA, WB-Re

Sequence: MGQGPRSPHKVGRRFPAGGKRGRGAKGSGRPLPGRKRQPWPPPDGRSEPAPDSHPHLSPEALGYFRRALSALKEAPETGEERDLMVHNIMKEVETQALALSTNRTGSEMLQELLGFSPLKPLCRVWAALRSNLRTVACHRCGVHVLQSALLQLPRLLGSAAEEEEEEEEEDGKDGPTETLEELVLGLAAEVCDDFLVYCGDTHGSFVVRTLLQVLGGTILESERARPRGSQSSEAQKTPAQECKPADFEVPETFLNRLQDLSSSFLKDIAVFITDKISSFCLQVALQVLHRKLPQFCAHLCNAVIGYLNTRGSSVDGSPLLLFLRDQTSSRLLEQVLLVLEPPRLQSLFEEHLQGQLQTLAAHPIANFPLQRLLDAVTTPELLSPVFEELSPVLEAVLAQGHPGVVIALVGACRRVGAYQAKVLQLLLEAFHCAEPSSRQVACVPLFATLMAYEVYYGLTEEEGAVPAEHQVAMAAARALGDVTVLGSLLLQHLLHFSTPGLVLRSLGALTGPQLLSLAQSPGRKLLLSLGSRTRS

Specifications

Accession Number AAH25332.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 161424
Molecular Weight (g/mol) 84.6kDa
Name C14orf21 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 ug
Immunogen MGQGPRSPHKVGRRFPAGGKRGRGAKGSGRPLPGRKRQPWPPPDGRSEPAPDSHPHLSPEALGYFRRALSALKEAPETGEERDLMVHNIMKEVETQALALSTNRTGSEMLQELLGFSPLKPLCRVWAALRSNLRTVACHRCGVHVLQSALLQLPRLLGSAAEEEEEEEEEDGKDGPTETLEELVLGLAAEVCDDFLVYCGDTHGSFVVRTLLQVLGGTILESERARPRGSQSSEAQKTPAQECKPADFEVPETFLNRLQDLSSSFLKDIAVFITDKISSFCLQVALQVLHRKLPQFCAHLCNAVIGYLNTRGSSVDGSPLLLFLRDQTSSRLLEQVLLVLEPPRLQSLFEEHLQGQLQTLAAHPIANFPLQRLLDAVTTPELLSPVFEELSPVLEAVLAQGHPGVVIALVGACRRVGAYQAKVLQLLLEAFHCAEPSSRQVACVPLFATLMAYEVYYGLTEEEGAVPAEHQVAMAAARALGDVTVLGSLLLQHLLHFSTPGLVLRSLGALTGPQLLSLAQSPGRKLLLSLGSRTRS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA2021
Common Name C14orf21
Gene Symbol C14orf21
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.