Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human C1qTNF5 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC3167R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3167R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3167R100 Supplier R&D Systems Supplier No. IC3167R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

C1qTNF5/CTRP5 Monoclonal specifically detects C1qTNF5/CTRP5 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen C1qTNF5/CTRP5
Applications Flow Cytometry
Classification Monoclonal
Clone 332923
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9BXJ0
Gene Alias C1q and tumor necrosis factor related protein 5, C1qTNF5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human C1qTNF5 Ser16-Ala243 (Gln44Arg) Accession # Q9BXJ0
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114902
Test Specificity Detects human C1qTNF5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) C1qTNF1 or rhC1qTNF4 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.