Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human C1qTNF9 ELISA Kit

Catalog No. p-7219713
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests

Catalog No. EH55RBX5

Supplier: Invitrogen™ EH55RBX5

Only null left
Add to Cart
Add to Cart

ELISA

Human C1qTNF9 quantitates human C1qTNF9 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human C1qTNF9.

Probable adipokine. Activates AMPK, AKT, and p44/42 MAPK signaling pathways
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 9130217G22Rik; AQL1; C1q and tumor necrosis factor related protein 9; C1qtnf9; C1QTNF9A; Ciqtnf9; Complement C1q and tumor necrosis factor-related protein 9; complement C1q and tumor necrosis factor-related protein 9 A; complement C1q tumor necrosis factor-related protein 9; complement C1q tumor necrosis factor-related protein 9 A; CTRP9; RGD1560876; UNQ6503/PRO21380
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 338872
Gene Symbol C1QTNF9
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target C1QTNF9
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.