Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human C5L2/GPR77 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10254R10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10254R10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10254R10 Supplier R&D Systems Supplier No. FAB10254R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

C5L2/GPR77 Monoclonal specifically detects C5L2/GPR77 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen C5L2/GPR77
Applications Flow Cytometry
Classification Monoclonal
Clone 650918
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9P296
Gene Alias C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77, GPR77
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human C5L2/GPR77, Met1-Val337, Accession # Q9P296
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27202
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.