Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human C5L2/GPR77 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10254N100UG
This item is not returnable.
View return policy
Description
C5L2/GPR77 Monoclonal specifically detects C5L2/GPR77 in Human samples. It is validated for Flow Cytometry.Specifications
C5L2/GPR77 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77, GPR77 | |
NS0 mouse myeloma cell line transfected with human C5L2/GPR77, Met1-Val337, Accession # Q9P296 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
650918 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9P296 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27202 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction