Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human C6orf206 Full-length ORF (NP_689945.2, 1 a.a. - 276 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Description
Radial spokes are regularly spaced along cilia, sperm, and flagella axonemes and have a multisubunit 'stalk' and 'head' that form a signal transduction scaffold between the central microtubule pair and dynein (see MIM 603297) arms. RSPH9 is predicted to be a component of the radial spoke head based on homology with proteins in the biflagellate alga Chlamydomonas reinhardtii and other ciliates (Castleman et al., 2009 [PubMed 19200523]).[supplied by OMIM]
Sequence: MDADSLLLSLELASGSGQGLSPDRRASLLTSLMLVKRDYRYDRVLFWGRILGLVADYYIAQGLSEDQLAPRKTLYSLNCTEWSLLPPATEEMVAQSSVVKGRFMGDPSYEYEHTELQKVNEGEKVFEEEIVVQIKEETRLVSVIDQIDKAVAIIPRGALFKTPFGPTHVNRTFEGLSLSEAKKLSSYFHFREPVELKNKTLLEKADLDPSLDFMDSLEHDIPKGSWSIQMERGNALVVLRSLLWPGLTFYHAPRTKNYGYVYVGTGEKNMDLPFML
Specifications
Specifications
Accession Number | NP_689945.2 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 221421 |
Molecular Weight (g/mol) | 57.7kDa |
Name | C6orf206 (Human) Recombinant Protein (P01) |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 25 μg |
Immunogen | MDADSLLLSLELASGSGQGLSPDRRASLLTSLMLVKRDYRYDRVLFWGRILGLVADYYIAQGLSEDQLAPRKTLYSLNCTEWSLLPPATEEMVAQSSVVKGRFMGDPSYEYEHTELQKVNEGEKVFEEEIVVQIKEETRLVSVIDQIDKAVAIIPRGALFKTPFGPTHVNRTFEGLSLSEAKKLSSYFHFREPVELKNKTLLEKADLDPSLDFMDSLEHDIPKGSWSIQMERGNALVVLRSLLWPGLTFYHAPRTKNYGYVYVGTGEKNMDLPFML |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction