Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CA125/MUC16 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5609G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5609G-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5609G-100 Supplier R&D Systems Supplier No. FAB5609G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CA125/MUC16
Applications Flow Cytometry
Classification Monoclonal
Clone 986808
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1μg/106 cells
Formulation Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. NP_078966.2
Gene Alias CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, MUC16, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CA125/MUC-16, Met13360-Gln14347 (Met13472Thr & Gln13957Lys), Accession # NP_078966.2
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 94025
Test Specificity Detects human CA125/MUC-16 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.