Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CA125/MUC16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $561.50
Specifications
Antigen | CA125/MUC16 |
---|---|
Clone | 606722R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB56092SP
![]() |
R&D Systems
MAB56092SP |
25 μg |
Each for $214.00
|
|
|||||
MAB56092100
![]() |
R&D Systems
MAB56092100 |
100 μg |
Each for $561.50
|
|
|||||
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for ELISA.Specifications
CA125/MUC16 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
NP_078966.2 | |
94025.0 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
606722R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, MUC16, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
Chinese hamster ovary cell line CHO-derived recombinant human CA125/MUC16, Met13360-Gln14347 (Met13472Thr & Gln13957Lys), Accession # NP_078966.2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CA125/MUC16 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title