Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CA125/MUC16 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB56091100
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
CA125/MUC16 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, MUC16, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
Chinese hamster ovary cell line CHO-derived recombinant human CA125/MUC-16, Met13360-Gln14347 (Met13472Thr & Gln13957Lys), Accession # NP_078966.2 | |
100 μg | |
94025 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
CyTOF, Flow Cytometry, Immunohistochemistry | |
986811 | |
Flow Cytometry 0.25μL/106 cells, Immunohistochemistry 5-25μg/mL, CyTOF-ready | |
NP_078966.2 | |
Mouse | |
Protein A or G purified from ascites | |
Primary | |
Detects human CA125/MUC-16 in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction