Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human CA19-9 ELISA Kit (Colorimetric)
SDP

Catalog No. NB599631KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB599631KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB599631KT Supplier Novus Biologicals™ Supplier No. NBP2599631KIT
Add to Cart
Add to Cart

Assay has high sensitivity and specificity for detection of human CA19-9

The Human CA19-9/Sialyl Lewis A ELISA Kit (Colorimetric) measures human CA19-9/Sialyl Lewis A in serum, plasma.

Specifications

Assay Range 10 U/ml to 150 U/ml
Assay Sensitivity 2.5 U/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of human CA19-9
Target Species Human
For Use With (Application) ELISA
Kit Contents Instruction manual, Substrate A, Stop Solution, Standard, Sample Diluent, Wash Buffer (20 x concentrate), Substrate B, HRP-conjugate, Adhesive Strip (For 96 wells), Assay plate
Quantity 1 Kit
Research Discipline Cancer
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym Sialyl Lewis (a), Sialyl Lewis(a), Sialyl Lewisa
Target CA19-9
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.