Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cadherin-11 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA17901T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA17901T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA17901T100 Supplier R&D Systems Supplier No. FAB17901T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cadherin-11 Monoclonal specifically detects Cadherin-11 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Cadherin-11
Applications Flow Cytometry
Classification Monoclonal
Clone 667039
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAA35622
Gene Alias CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, Cadherin11, CDH11, CDHOB, OB, OB-cadherin, Osteoblast cadherin
Gene Symbols CDH11
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cadherin-11 Phe23-Thr617 Accession # AAA35622
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1009
Test Specificity Detects human Cadherin-11 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Cadherin-4, -6, -8, -12, -13, -17, rhE-Cadherin, rhN-Cadherin, rhP-Cadherin, or rhVE-Cadherin is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.