Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cadherin-11 ELISA Kit

Catalog No. EH61RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH61RBX5 5 x 96 Tests
EH61RBX10 10 x 96 Tests
EH61RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH61RBX5 Supplier Invitrogen™ Supplier No. EH61RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human Cadherin-11 quantitates human Cadherin-11 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cadherin-11.

This gene encodes a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. All cadherins are single-pass transmembrane proteins with a variable number of 110 amino acid extracellular cadherin (EC) domains. Classical cadherins contain five large N-terminal EC domains, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Based on amino acid alignment, classical cadherins are divided into type I and type II subgroups. Type I cadherins, including cadherins E (epithelial), N (neural), P (placental), and R (retinal) cadherin differ from type I cadherins in their specific amino acid sequences. Type II cadherins include human cadherin-5, -6, -8, -11, and -12. Type II cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Cadherin-11 (OB-Cadherin) is a marker of the loosely connected and migratory cellular elements of the mesenchyme. Strong expression of Cadherin-11 has been noted in brain, spinal cord, bone marrow and bone cells, and is regulated in human endometrial glandular epithelial and stromal cells. Expression of this particular cadherin in osteoblastic cell lines, and its upregulation during differentiation, suggests a specific function in bone development and maintenance. A variant Cadherin-11 gene (variant form) encodes a truncated protein with an unusual cytoplasmic domain not present in other cadherins. The variant protein is generated by alternative splicing, utilizing a 179 bp intronic sequence. Recently, Cadherin-11 mRNA and protein were found to be expressed in most invasive breast cancer cell lines tested but not in non-invasive cell lines, suggesting that Cadherin-11 may be a marker for aggressive, invasive tumor subsets.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P55287
Assay Range 0.8-200 ng/mL
Assay Sensitivity 0.8 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CAD11; Cad-11; Cadherin; cadherin 11; cadherin 11, type 2, OB-cadherin (osteoblast); cadherin-11; Cdh11; CDHOB; OB; OB Cadherin; OB-cadherin; OSF-4; Osteoblast cadherin; osteoblast-cadherinCAD11; Cad-11; Cadherin; cadherin 11; cadherin 11, type 2, OB-cadherin (osteoblast); cadherin-11; Cdh11; CDHOB; OB; OB Cadherin; OB-cadherin; OSF-4; Osteoblast cadherin; osteoblast-cadherin;
Gene ID (Entrez) 1009
Gene Symbol CDH11
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CDH11
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.