Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cadherin-12 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB2240G100UG
This item is not returnable.
View return policy
Description
Cadherin-12 Monoclonal specifically detects Cadherin-12 in Human samples. It is validated for Flow Cytometry.Specifications
Cadherin-12 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P55289 | |
CDH12 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-12 Met1-Pro609 Accession # P55289 | |
RUO | |
1010 | |
Human | |
IgG1 |
Flow Cytometry | |
343621 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
BR-cadherin, cadherin 12, type 2 (N-cadherin 2), cadherin-12, Cadherin12, CDH12, CDHB, FLJ34857, N-Cadherin 2, Neural type cadherin 2, neural type, 2, neuronal cadherin 2 | |
Rat | |
100 μg | |
Primary | |
Detects human Cadherin-12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cadherin-8, -11, -13, -17, rhE-Cadherin, rhP-Cadherin, rhR-Cadherin, or rhVR-Cadherin is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction