Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cadherin-17 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1032SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1032SP 25 μg
MAB1032100 100 μg
MAB1032500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1032SP Supplier R&D Systems Supplier No. MAB1032SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Cadherin-17 Monoclonal specifically detects Cadherin-17 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Cadherin-17
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 141713
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA58231
Gene Alias cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, Cadherin17, CDH16, CDH17, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024
Gene Symbols CDH17
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cadherin-17 Gln23-Met787 Accession # CAA58231
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1015
Test Specificity Detects human Cadherin-17 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Cadherin-8, rhE-Cadherin, rhP-Cadherin, or rhVE-Cadherin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.