Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cadherin-4/R-Cadherin Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Cadherin-4/R-Cadherin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Cadherin-4/R-Cadherin Polyclonal specifically detects Cadherin-4/R-Cadherin in Human samples. It is validated for Western Blot.Specifications
Cadherin-4/R-Cadherin | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, Cadherin4, CDH4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
CDH4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P55283 | |
1002 | |
Mouse myeloma cell line NS0-derived recombinant human Cadherin-4/R-Cadherin His21-Ala734 (Lys347Trp) Accession # P55283 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Cadherin-4/R-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) N-Cadherin is observed and less than 1% cross-reactivity with rhCadherin-8, rhCadherin-11, rhCadherin-12, rhCadherin-17, rhE-Cadherin, rhP-Cadherin, and rhVE-Cadherin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title