Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Calcium Activated Nucleotidase 1/CANT1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6720
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6720 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6720 Supplier R&D Systems Supplier No. AF6720
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Calcium Activated Nucleotidase 1/CANT1 Polyclonal specifically detects Calcium Activated Nucleotidase 1/CANT1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Calcium Activated Nucleotidase 1/CANT1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q8WVQ1
Gene Alias Apyrase homolog, calcium activated nucleotidase 1, DBQD, EC 3.6.1.6, Putative MAPK-activating protein PM09, Putative NF-kappa-B-activating protein 107, SCAN1, SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase, SHAPY, SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification, soluble Ca-activated nucleotidase, isozyme 1, soluble calcium-activated nucleotidase 1, soluble calcium-activated nucleotidase SCAN-1
Gene Symbols CANT1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Calcium Activated Nucleotidease 1/CANT1 Gly80-Ile401 Accession # Q8WVQ1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124583
Test Specificity Detects human Calcium Activated Nucleotidease 1/CANT1 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.