Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Calcium-sensing R/CaSR Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | Calcium-sensing R/CaSR |
---|---|
Clone | 611825 |
Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9608SP
![]() |
R&D Systems
MAB9608SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9608100
![]() |
R&D Systems
MAB9608100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Calcium-sensing R/CaSR Monoclonal specifically detects Calcium-sensing R/CaSR in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Calcium-sensing R/CaSR | |
Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
P41180-1 | |
846 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
611825 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2A, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441 | |
Mouse myeloma cell line NS0-derived transfected with recombinant human Calcium-sensing R/CaSR protein, Tyr20-Lys601, Accession # P41180-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Calcium-sensing R/CaSR protein in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title