Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Calponin 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7900SP
This item is not returnable.
View return policy
Description
Calponin 1 Monoclonal specifically detects Calponin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
Calponin 1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P51911 | |
CNN1 | |
E. coli-derived recombinant human Calponin 1 Ser2-Ala297 Accession # P51911 | |
25 μg | |
Primary | |
Detects human Calponin 1 in ELISAs. Detects human, mouse and rat Calponin 1 in Western Blots. In direct ELISAs, no cross-reactivity with recombinant human Calponin 3 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
836701 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, Calponin H1, smooth muscle, calponins, basic, CNN1, Sm-Calp, SMCC, SMCCcalponin-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1264 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction